Isochinolin-7-amin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 7-Aminoisoquinoline, Isoquinolin-7-amine

AS1281G1 AS1281G5
ACTIAS1281G1EA 0 CHF
ACTIAS1281G1 ACTIAS1281G5
Isochinolin-7-amin
Isochinolin-7-amin
Formel: C₉H₈N₂
MW: 144,1700 g/mol
MDL Number: MFCD08669932
CAS-Nummer: 23707-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR