Isochinolin-3-amin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Isoquinolin-3-amine, 3-Aminoisoquinoline

AS31607G1 AS31607G5
ACTIAS31607G1EA 0 CHF
ACTIAS31607G1 ACTIAS31607G5
Isochinolin-3-amin
Isochinolin-3-amin
Formel: C₉H₈N₂
MW: 144,1700 g/mol
MDL Number: MFCD00102190
CAS-Nummer: 25475-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR