(±)-tert-Butyl-2-brombutyrat

Lieferant: ACTIVATE SCIENTIFIC

AS23098G25 AS23098G5
ACTIAS23098G25EA 0 CHF
ACTIAS23098G25 ACTIAS23098G5
(±)-tert-Butyl-2-brombutyrat
(±)-tert-Butyl-2-brombutyrat
Formel: C₈H₁₅BrO₂
MDL Number: MFCD00209634
CAS-Nummer: 24457-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR