α-Brom-5-chlor-2-nitrotoluol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-(Bromomethyl)-4-chloro-1-nitrobenzene, (2-Bromomethyl)-4-chloro-1-nitrobenzene
AS2722G1
AS2722G5
ACTIAS2722G1EA
50
CHF
ACTIAS2722G1
ACTIAS2722G5
α-Brom-5-chlor-2-nitrotoluol
α-Brom-5-chlor-2-nitrotoluol
Formel:
C₇H₅BrClNO₂ MW: 250,4771 g/mol |
MDL Number:
MFCD11848697 CAS-Nummer: 31577-25-0 EINECS: 260-179-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...