2-Amino-5-brom-4-methyl-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33973G1 AS33973G10 AS33973G5
ACTIAS33973G1EA 0 CHF
ACTIAS33973G1 ACTIAS33973G10 ACTIAS33973G5
2-Amino-5-brom-4-methyl-3-nitropyridin
2-Amino-5-bromo-4-methyl-3-nitropyridine
Formel: C₆H₆BrN₃O₂
MDL Number: MFCD00092197
CAS-Nummer: 100367-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR