N-Phenylisonicotinamid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: N-Phenyl isonicotinicamide, N-Phenyl-4-pyridinecarboxamide

AS35411G1 AS35411G5
ACTIAS35411G1EA 0 CHF
ACTIAS35411G1 ACTIAS35411G5
N-Phenylisonicotinamid
N-Phenylisonicotinamid
Formel: C₁₂H₁₀N₂O
MW: 198,2200 g/mol
MDL Number: MFCD00459246
CAS-Nummer: 3034-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR