5-Brom-2-(dimethylamino)pyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-N,N-dimethylpyridin-2-amine

AS50604G1 AS50604G5
ACTIAS50604G1EA 0 CHF
ACTIAS50604G1 ACTIAS50604G5
5-Brom-2-(dimethylamino)pyridin
5-Brom-2-(dimethylamino)pyridin
Formel: C₇H₉BrN₂
MW: 201.0600 g/mol
MDL Number: MFCD00099486
CAS-Nummer: 26163-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR