3-Brom-5-chlor-2-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35273G1 AS35273G5
ACTIAS35273G1EA 0 CHF
ACTIAS35273G1 ACTIAS35273G5
3-Brom-5-chlor-2-hydroxypyridin
3-Bromo-5-chloro-2-hydroxypyridine
Formel: C₅H₃BrClNO
MW: 208.4400 g/mol
MDL Number: MFCD01764522
CAS-Nummer: 137628-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR