Isochinolin-6-carbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS1291G1 AS1291G5
ACTIAS1291G1EA 0 CHF
ACTIAS1291G1 ACTIAS1291G5
Isochinolin-6-carbonitril
Isochinolin-6-carbonitril
Formel: C₁₀H₆N₂
MW: 154,1680 g/mol
MDL Number: MFCD09991660
CAS-Nummer: 106778-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR