Isochinolin-5-carbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS1290G1 AS1290G5
ACTIAS1290G1EA 0 CHF
ACTIAS1290G1 ACTIAS1290G5
Isochinolin-5-carbonitril
Isochinolin-5-carbonitril
Formel: C₁₀H₆N₂
MDL Number: MFCD03109890
CAS-Nummer: 27655-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR