Isochinolin-4-carbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS1289G1 AS1289G5
ACTIAS1289G1EA 0 CHF
ACTIAS1289G1 ACTIAS1289G5
Isochinolin-4-carbonitril
Isochinolin-4-carbonitril
Formel: C₁₀H₆N₂
MW: 154,1700 g/mol
MDL Number: MFCD00957039
CAS-Nummer: 34846-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR