Isochinolin-3-carbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Isoquinolinecarbonitrile

AS36002G1 AS36002G5
ACTIAS36002G1EA 0 CHF
ACTIAS36002G1 ACTIAS36002G5
Isochinolin-3-carbonitril
Isochinolin-3-carbonitril
Formel: C₁₀H₆N₂
MW: 154,17 g/mol
MDL Number: MFCD00075136
CAS-Nummer: 26947-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR