Isochinolin-1-carbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Isoquinolinecarbonitrile

AS35451G1 AS35451G5
ACTIAS35451G1EA 0 CHF
ACTIAS35451G1 ACTIAS35451G5
Isochinolin-1-carbonitril
Isochinolin-1-carbonitril
Formel: C₁₀H₆N₂
MDL Number: MFCD00134166
CAS-Nummer: 1198-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR