Isochinolin-8-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS1308G1 AS1308G5
ACTIAS1308G1EA 0 CHF
ACTIAS1308G1 ACTIAS1308G5
Isochinolin-8-carbaldehyd
Isochinolin-8-carbaldehyd
Formel: C₁₀H₇NO
MDL Number: MFCD06227440
CAS-Nummer: 787615-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR