Isochinolin-5-carbaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Isoquinoline-5-carboxaldehyde
AS1433G1
AS1433G5
ACTIAS1433G1EA
0
CHF
ACTIAS1433G1
ACTIAS1433G5
Isochinolin-5-carbaldehyd
Isochinolin-5-carbaldehyd
Formel:
C₁₀H₇NO MW: 157,17 g/mol |
MDL Number:
MFCD03412483 CAS-Nummer: 80278-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...