Isochinolin-4-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS1305G1 AS1305G5
ACTIAS1305G1EA 0 CHF
ACTIAS1305G1 ACTIAS1305G5
Isochinolin-4-carbaldehyd
Isochinolin-4-carbaldehyd
Formel: C₁₀H₇NO
MDL Number: MFCD00829440
CAS-Nummer: 22960-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR