6-Brom-3-isochinolinol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Bromo-3-hydroxyisoquinoline

AS1985G1 AS1985G5
ACTIAS1985G1EA 0 CHF
ACTIAS1985G1 ACTIAS1985G5
6-Brom-3-isochinolinol
6-Brom-3-isochinolinol
Formel: C₉H₆BrNO
MW: 224,0500 g/mol
MDL Number: MFCD11110423
CAS-Nummer: 1031927-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR