4-Brom-1-methoxyisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS30787G1 AS30787G5
ACTIAS30787G1EA 0 CHF
ACTIAS30787G1 ACTIAS30787G5
4-Brom-1-methoxyisochinolin
4-Bromo-1-methoxyisoquinoline
Formel: C₁₀H₈BrNO
MW: 238,0800 g/mol
MDL Number: MFCD09999134
CAS-Nummer: 746668-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR