1-Amino-4-bromisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS11233G1 AS11233G5
ACTIAS11233G1EA 0 CHF
ACTIAS11233G1 ACTIAS11233G5
1-Amino-4-bromisochinolin
1-Amino-4-bromoisoquinoline
Formel: C₉H₇BrN₂
MDL Number: MFCD05155261
CAS-Nummer: 55270-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR