1-Cyclohexenylboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Cyclohexeneboronic acid pinacol ester, 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Cyclohexene-1-boronic acid pinacol ester

AS33090G1 AS33090G5
ACTIAS33090G1EA 0 CHF
ACTIAS33090G1 ACTIAS33090G5
1-Cyclohexenylboronsäurepinakolester
1-Cyclohexenylboronsäurepinakolester
Formel: C₁₂H₂₁BO₂
MW: 208.1049 g/mol
MDL Number: MFCD05663845
CAS-Nummer: 141091-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR