6-Chlor-5-nitrochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS31430G1 AS31430G5
ACTIAS31430G1EA 0 CHF
ACTIAS31430G1 ACTIAS31430G5
6-Chlor-5-nitrochinolin
6-Chloro-5-nitroquinoline
Formel: C₉H₅ClN₂O₂
MDL Number: MFCD00093616
CAS-Nummer: 86984-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR