4-Chlor-3-nitrochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS30597G1 AS30597G5
ACTIAS30597G1EA 0 CHF
ACTIAS30597G1 ACTIAS30597G5
4-Chlor-3-nitrochinolin
4-Chloro-3-nitroquinoline
Formel: C₉H₅ClN₂O₂
MW: 208.6012 g/mol
MDL Number: MFCD07369246
CAS-Nummer: 39061-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR