5-Amino-6-chlorchinolin
Lieferant: ACTIVATE SCIENTIFIC
AS35642G1
AS35642G5
ACTIAS35642G1EA
0
CHF
ACTIAS35642G1
ACTIAS35642G5
5-Amino-6-chlorchinolin
5-Amino-6-chloroquinoline
Formel:
C₉H₇ClN₂ MW: 178,6200 g/mol |
MDL Number:
MFCD02947117 CAS-Nummer: 341010-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...