4-Amino-6-bromchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS22272G1 AS22272G5
ACTIAS22272G1EA 173 CHF
ACTIAS22272G1 ACTIAS22272G5
4-Amino-6-bromchinolin
4-Amino-6-bromoquinoline
Formel: C₉H₇BrN₂
MW: 223,0693 g/mol
MDL Number: MFCD03407953
CAS-Nummer: 65340-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR