4-Amino-3-bromchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS30588G1 AS30588G5
ACTIAS30588G1EA 0 CHF
ACTIAS30588G1 ACTIAS30588G5
4-Amino-3-bromchinolin
4-Amino-3-bromoquinoline
Formel: C₉H₇BrN₂
MW: 223,07 g/mol
MDL Number: MFCD00234502
CAS-Nummer: 36825-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR