8-Brom-2-chlorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS13040G1 AS13040G5
ACTIAS13040G1EA 0 CHF
ACTIAS13040G1 ACTIAS13040G5
8-Brom-2-chlorchinolin
8-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
MDL Number: MFCD08741410
CAS-Nummer: 163485-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR