7-Brom-4-chlorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS2473G1 AS2473G5
ACTIAS2473G1EA 0 CHF
ACTIAS2473G1 ACTIAS2473G5
7-Brom-4-chlorchinolin
7-Bromo-4-chloroquinoline
Formel: C₉H₅BrClN
MDL Number: MFCD02684221
CAS-Nummer: 75090-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR