4-Brom-7-chlorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS33475G1 AS33475G5
ACTIAS33475G1EA 0 CHF
ACTIAS33475G1 ACTIAS33475G5
4-Brom-7-chlorchinolin
4-Bromo-7-chloroquinoline
Formel: C₉H₅BrClN
MW: 242,5000 g/mol
MDL Number: MFCD07700232
CAS-Nummer: 98519-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR