3-Brom-2-chlorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS1605G1 AS1605G5
ACTIAS1605G1EA 0 CHF
ACTIAS1605G1 ACTIAS1605G5
3-Brom-2-chlorchinolin
3-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
MDL Number: MFCD08457743
CAS-Nummer: 101870-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR