4-Benzyloxy-3-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Benzyloxy-3-fluorobenzeneboronic acid

AS50018G1 AS50018G5
ACTIAS50018G1EA 0 CHF
ACTIAS50018G1 ACTIAS50018G5
4-Benzyloxy-3-fluorphenylboronsäure
4-Benzyloxy-3-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
MW: 246,04 g/mol
MDL Number: MFCD00994627
CAS-Nummer: 133057-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR