4-Benzyloxy-2-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Benzyloxy-2-fluorobenzeneboronic acid, (4-(Benzyloxy)-2-fluorophenyl)boronic acid

AS50227G1 AS50227G5
ACTIAS50227G1EA 0 CHF
ACTIAS50227G1 ACTIAS50227G5
4-Benzyloxy-2-fluorphenylboronsäure
4-Benzyloxy-2-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
MW: 246,04 g/mol
MDL Number: MFCD02093064
CAS-Nummer: 166744-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR