4-Benzyloxy-2,6-difluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-(Benzyloxy)-2,6-difluorophenyl)boronic acid

AS36131G5 AS36131G1
ACTIAS36131G5EA 0 CHF
ACTIAS36131G5 ACTIAS36131G1
4-Benzyloxy-2,6-difluorphenylboronsäure
4-Benzyloxy-2,6-difluorphenylboronsäure
Formel: C₁₃H₁₁BF₂O₃
MDL Number: MFCD12026734
CAS-Nummer: 156635-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR