3-(Cyclopropylmethoxy)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (3-(Cyclopropylmethoxy)phenyl)boronic acid

AS1664G1 AS1664G5
ACTIAS1664G1EA 0 CHF
ACTIAS1664G1 ACTIAS1664G5
3-(Cyclopropylmethoxy)phenylboronsäure
3-(Cyclopropylmethoxy)phenylboronsäure
Formel: C₁₀H₁₃BO₃
MW: 192,200 g/mol
MDL Number: MFCD04039016
CAS-Nummer: 411229-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR