4-Cyclopropylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Cyclopropylphenyl)boronic acid

AS15242G1 AS15242G5
ACTIAS15242G1EA 0 CHF
ACTIAS15242G1 ACTIAS15242G5
4-Cyclopropylphenylboronsäure
4-Cyclopropylphenylboronsäure
Formel: C₉H₁₁BO₂
MW: 161.9934 g/mol
MDL Number: MFCD01309064
CAS-Nummer: 302333-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR