2,6-Dimethyl-4-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Hydroxy-2,6-dimethylbenzonitrile

AS32422G1 AS32422G5
ACTIAS32422G1EA 0 CHF
ACTIAS32422G1 ACTIAS32422G5
2,6-Dimethyl-4-hydroxybenzonitril
2,6-Dimethyl-4-hydroxybenzonitril
Formel: C₉H₉NO
MW: 147,1700 g/mol
MDL Number: MFCD00238591
CAS-Nummer: 58537-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR