4-Brom-3,5-dimethylbenzamid

Lieferant: ACTIVATE SCIENTIFIC

AS36454G1 AS36454G5
ACTIAS36454G1EA 0 CHF
ACTIAS36454G1 ACTIAS36454G5
4-Brom-3,5-dimethylbenzamid
4-Brom-3,5-dimethylbenzamid
Formel: C₉H₁₀BrNO
MW: 228,0900 g/mol
MDL Number: MFCD07369913
CAS-Nummer: 864825-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR