3-Methoxybenzamidin Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

AS30168G1 AS30168G5
ACTIAS30168G1EA 0 CHF
ACTIAS30168G1 ACTIAS30168G5
3-Methoxybenzamidin Hydrochlorid
3-Methoxybenzamidin Hydrochlorid
Formel: C₈H₁₁ClN₂O
MW: 186,6400 g/mol
MDL Number: MFCD04114430
CAS-Nummer: 26113-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR