3-(Aminomethyl)benzoesäure Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

AS33912G1 AS33912G5
ACTIAS33912G1EA 0 CHF
ACTIAS33912G1 ACTIAS33912G5
3-(Aminomethyl)benzoesäure Hydrochlorid
3-(Aminomethyl)benzoesäure Hydrochlorid
Formel: C₈H₁₀ClNO₂
MDL Number: MFCD03791117
CAS-Nummer: 876-03-9
EINECS: 239-212-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR