tert-Butyl-4-(chlormethyl)benzoat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
tert-Butyl 4-(chloromethyl)benzoate
AS35381G1
AS35381G10
AS35381G5
ACTIAS35381G1EA
0
CHF
ACTIAS35381G1
ACTIAS35381G10
ACTIAS35381G5
tert-Butyl-4-(chlormethyl)benzoat
tert-Butyl-4-(chlormethyl)benzoat
Formel:
C₁₂H₁₅ClO₂ MW: 226,7000 g/mol |
MDL Number:
MFCD15142807 CAS-Nummer: 121579-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...