4-(Brommethyl)benzolsulfonamid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Bromomethylbenzenesulphonamide
AS30465G1
AS30465G5
ACTIAS30465G1EA
0
CHF
ACTIAS30465G1
ACTIAS30465G5
4-(Brommethyl)benzolsulfonamid
4-(Brommethyl)benzolsulfonamid
Formel:
C₇H₈BrNO₂S MW: 250,1100 g/mol |
MDL Number:
MFCD08752696 CAS-Nummer: 40724-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...