3-(Brommethyl)benzolsulfonamid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromomethylbenzenesulphonamide

AS36717G1 AS36717G5
ACTIAS36717G1EA 309 CHF
ACTIAS36717G1 ACTIAS36717G5
3-(Brommethyl)benzolsulfonamid
3-(Brommethyl)benzolsulfonamid
Formel: C₇H₈BrNO₂S
MW: 250,1100 g/mol
MDL Number: MFCD09743699
CAS-Nummer: 220798-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR