2-Methoxybenzolsulfonamid

Lieferant: ACTIVATE SCIENTIFIC

AS15204G1 AS15204G5
ACTIAS15204G1EA 94 CHF
ACTIAS15204G1 ACTIAS15204G5
2-Methoxybenzolsulfonamid
2-Methoxybenzolsulfonamid
Formel: C₇H₉NO₃S
MDL Number: MFCD06496492
CAS-Nummer: 52960-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR