1-(Methylsulfonyl)-3-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS22814G1 AS22814G25 AS22814G5
ACTIAS22814G1EA 150 CHF
ACTIAS22814G1 ACTIAS22814G25 ACTIAS22814G5
1-(Methylsulfonyl)-3-nitrobenzol
1-(Methylsulphonyl)-3-nitrobenzene
Formel: C₇H₇NO₄S
MW: 201,1998 g/mol
MDL Number: MFCD00091590
CAS-Nummer: 2976-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR