4-(Difluormethoxy)benzolsulfonamid

Lieferant: ACTIVATE SCIENTIFIC

AS24709G1 AS24709G25 AS24709G5
ACTIAS24709G1EA 78 CHF
ACTIAS24709G1 ACTIAS24709G25 ACTIAS24709G5
4-(Difluormethoxy)benzolsulfonamid
4-(Difluormethoxy)benzolsulfonamid
Formel: C₇H₇F₂NO₃S
MW: 223.2000 g/mol
MDL Number: MFCD03407973
CAS-Nummer: 874781-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR