4-Fluor-3-nitrobenzolsulfonamid

Lieferant: ACTIVATE SCIENTIFIC

AS21570G1 AS21570G25 AS21570G5
ACTIAS21570G1EA 23 CHF
ACTIAS21570G1 ACTIAS21570G25 ACTIAS21570G5
4-Fluor-3-nitrobenzolsulfonamid
4-Fluor-3-nitrobenzolsulfonamid
Formel: C₆H₅FN₂O₄S
MW: 220,18 g/mol
MDL Number: MFCD08703185
CAS-Nummer: 406233-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR