4-Cyan-3-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Cyano-3-fluorobenzeneboronic acid

AS34949G1 AS34949G5
ACTIAS34949G1EA 0 CHF
ACTIAS34949G1 ACTIAS34949G5
4-Cyan-3-fluorphenylboronsäure
4-Cyan-3-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
MW: 164,93 g/mol
MDL Number: MFCD03411549
CAS-Nummer: 843663-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR