3-Cyan-4-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Cyano-4-fluorobenzeneboronic acid

AS35393G1 AS35393G5
ACTIAS35393G1EA 0 CHF
ACTIAS35393G1 ACTIAS35393G5
3-Cyan-4-fluorphenylboronsäure
3-Cyan-4-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
MW: 164,93 g/mol
MDL Number: MFCD03095130
CAS-Nummer: 214210-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR