4-(Dimethylamino)benzamid
Lieferant: ACTIVATE SCIENTIFIC
AS36306G1
AS36306G5
ACTIAS36306G1EA
0
CHF
ACTIAS36306G1
ACTIAS36306G5
4-(Dimethylamino)benzamid
4-(Dimethylamino)benzamid
Formel:
C₉H₁₂N₂O MW: 164,2000 g/mol |
MDL Number:
MFCD01216782 CAS-Nummer: 6083-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...