4-Amino-2-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS36325G1 AS36325G5
ACTIAS36325G1EA 0 CHF
ACTIAS36325G1 ACTIAS36325G5
4-Amino-2-methoxybenzonitril
4-Amino-2-methoxybenzonitril
Formel: C₈H₈N₂O
MDL Number: MFCD09027303
CAS-Nummer: 7251-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR