4-Amino-3-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Cyano-2-methoxy-aniline

AS34361G1 AS34361G5
ACTIAS34361G1EA 0 CHF
ACTIAS34361G1 ACTIAS34361G5
4-Amino-3-methoxybenzonitril
4-Amino-3-methoxybenzonitril
Formel: C₈H₈N₂O
MDL Number: MFCD13250177
CAS-Nummer: 177476-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR